Assay Details
| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039176.2 | NP_001034265.1 | 3 - 4 | 386 | 71 | |
| NM_019422.3 | NP_062295.1 | 3 - 4 | 379 | 71 | ||
| GenBank mRNA | AF170907.1 | - | 3 - 4 | 372 | 71 | |
| AK135348.1 | - | 3 - 4 | 354 | 71 | ||
| AK156003.1 | - | 3 - 4 | 1980 | 71 | ||
| AK171217.1 | - | 3 - 4 | 365 | 71 | ||
| BC006735.1 | - | 3 - 4 | 386 | 3156004 | 71 | |
| BY773043.1 | - | 3 - 4 | 340 | 71 | ||
| CN455861.1 | - | 1 - 2 | 123 | 71 | ||
| CX199666.1 | - | 3 - 4 | 352 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elovl1
Genomic Map