Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_001204165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_001204165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_001204166.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_001204166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_019688.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
NM_019688.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008572.mm.1 |
TC0200001049.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_001204165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_001204165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_001204166.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_001204166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_019688.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
NM_019688.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008573.mm.1 |
TC0200001049.mm.1 |
MTA TC |