Assay Details
Gene Symbol: Crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019922.2 | NP_064306.2 | 4 - 5 | 1037 | 98 | |
GenBank mRNA | AJ006469.1 | - | 4 - 5 | 995 | 98 | |
AK017797.1 | - | 4 - 5 | 1000 | 98 | ||
AK047506.1 | - | 4 - 5 | 993 | 98 | ||
AK203540.1 | - | 2 - 3 | 177 | 98 | ||
BC049890.1 | - | 4 - 5 | 974 | 6306169 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
