Assay Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019922.2 | NP_064306.2 | 5 - 6 | 1178 | 62 | |
| GenBank mRNA | AJ006469.1 | - | 5 - 6 | 1136 | 62 | |
| AK007624.1 | - | 5 - 6 | 899 | 62 | ||
| AK017797.1 | - | 5 - 6 | 1141 | 62 | ||
| AK047506.1 | - | 5 - 6 | 1134 | 62 | ||
| AK192515.1 | - | 1 - 2 | 45 | 62 | ||
| AK195299.1 | - | 1 - 2 | 45 | 62 | ||
| AK198132.1 | - | 1 - 2 | 45 | 62 | ||
| AK199931.1 | - | 1 - 2 | 63 | |||
| BC049890.1 | - | 5 - 6 | 1115 | 6306169 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844285 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844414 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844378 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844377 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844286 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844413 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395950 | AK006738.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395950 | XM_001479989.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64635 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159944 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161750 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26844412 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161750 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159944 | - | - |
| Whole Mouse Genome G4122A | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | BC049890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AJ006469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK007624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK017797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P201721 | AK047506.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130289 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159944 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07716420 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07724405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07716421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07716416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07757014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844377 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844414 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844378 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844285 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844286 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26844380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26844390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07863165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07604104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07486421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07569712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07858156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07715769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07725608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07802662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07725609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07802657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07802656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07715772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26844415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597414 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448592_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597413 | - | - |
| Mouse Genome 430 2.0 Array | 1448592_at | - | - |
| Mouse Genome 430 2.0 Array | 1448592_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56693_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56693_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900026821.mm.1 | NM_019922.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026821.mm.1 | NM_019922.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900026821.mm.1 | TC0900003168.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448592_PM_at | - | - |
| Mouse Expression Set 430 | 1448592_at | - | - |
| Murine Genome U74v2 Set | 103817_at | - | - |
| Murine Genome U74v2 Set | 103817_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448592_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448592_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448592_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448592_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 761914 | - | - |
| ABI Mouse Genome Survey Microarray | 925891 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3990725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223997 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223997 | - | - |
| mouse-6 v1.1 (Illumina) | scl35279.4.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35279.4.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2877541 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223997 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2877541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223997 | - | - |

Crtap
Genomic Map