Assay Details
Gene Symbol: Chst5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019950.2 | NP_064334.1 | 1 - 2 | 70 | 109 | |
GenBank mRNA | AF176840.1 | - | 1 - 2 | 93 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF176840.1, NM_019950.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
N-acetylglucosamine metabolic process
sulfur compound metabolic process
keratan sulfate biosynthetic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
carbohydrate metabolic process
N-acetylglucosamine metabolic process
sulfur compound metabolic process
keratan sulfate biosynthetic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170843 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170773 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170766 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170840 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170768 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170772 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170771 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170842 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170770 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054027 | NM_019950.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479790 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46216 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759388 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623028 | AB017027.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623028 | BC087730.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623028 | NM_018766.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306867 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51911 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126308 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115438 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129372 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054027 | NM_019950.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299107 | AF176840.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170827 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170828 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26170852 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06643319 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06643314 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06643336 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129372 | - | - |
Whole Mouse Genome G4122A | A_51_P299107 | AF176840.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105325 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129372 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06643319 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06967271 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06967275 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06967268 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06967264 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170843 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170767 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170768 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170770 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170842 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170840 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170771 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170772 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170766 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26170773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06963545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06803802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06834806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170823 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06700913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06967271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06848429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06841409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26170827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581916 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581915 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419320_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581914 | - | - |
Mouse Genome 430 2.0 Array | 1419320_at | - | - |
Mouse Genome 430 2.0 Array | 1419320_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56773_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56773_at | - | - |
Mouse Expression Set 430 | 1419320_PM_at | - | - |
Mouse Expression Set 430 | 1419320_at | - | - |
Mouse Genome 430A 2.0 Array | 1419320_at | - | - |
Mouse Genome 430A 2.0 Array | 1419320_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419320_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419320_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 635209 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6900600 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6900600 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900600 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665754 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2665754 | - | - |
mouse-6 v1.1 (Illumina) | scl34303.2_339-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900600 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34303.2_339-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900600 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2665754 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665754 | - | - |
