Assay Details
Gene Symbol: Dnajb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019965.2 | NP_064349.2 | 1 - 2 | 220 | 76 | |
GenBank mRNA | AB028860.1 | - | 1 - 2 | 193 | 76 | |
AK038149.1 | - | 1 - 2 | 247 | 76 | ||
AK082546.1 | - | 1 - 2 | 170 | 76 | ||
AK089427.1 | - | 1 - 2 | 189 | 76 | ||
AK138495.1 | - | 1 - 2 | 150 | 76 | ||
BC034162.1 | - | 1 - 2 | 220 | 5043868 | 76 |
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC006184.2 | - | 215 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
