Assay Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019975.3 | NP_064359.2 | 7 - 8 | 651 | 73 | |
| GenBank mRNA | AJ132139.1 | - | 7 - 8 | 649 | 73 | |
| AK005505.1 | - | 7 - 8 | 638 | 73 | ||
| AK041686.1 | - | 7 - 8 | 647 | 73 | ||
| AK050078.1 | - | 7 - 8 | 651 | 73 | ||
| AK161488.1 | - | 631 | 73 | |||
| BC021360.1 | - | 7 - 8 | 582 | 5011451 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hacl1
Genomic Map