Assay Details
| Gene Symbol: Hist1h1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020034.2 | NP_064418.1 | 1 | 689 | 66 | |
| GenBank mRNA | BC114579.1 | - | 1 | 617 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874556 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874571 | - | - |
| Whole Mouse Genome G4122A | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270949 | NM_020034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270949 | AK020117.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270949 | BC114579.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126244 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10456933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10456945 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10704466 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10456936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10456940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10760751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10456926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10456950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10760749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10456944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10456940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10456955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10760756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28874523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408082 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408081 | - | - |
| Murine Genome U74v2 Set | 99803_at | - | - |
| Murine Genome U74v2 Set | 99803_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 455012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1410592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1410592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001410592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664593 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664593 | - | - |
| mouse-6 v1.1 (Illumina) | scl44228.1.74_105-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1410592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44228.1.74_105-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664593 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664593 | - | - |

Hist1h1b
Genomic Map