Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700010858.mm.1 |
NM_020578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010858.mm.1 |
NM_020578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010858.mm.1 |
TC1700001154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010859.mm.1 |
NM_020578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010859.mm.1 |
NM_020578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010859.mm.1 |
TC1700001154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010861.mm.1 |
NM_020578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010861.mm.1 |
NM_020578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010861.mm.1 |
TC1700001154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010857.mm.1 |
NM_020578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010857.mm.1 |
NM_020578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010857.mm.1 |
TC1700001154.mm.1 |
MTA TC |