Assay Details
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020578.3 | NP_065603.2 | 4 - 5 | 1324 | 87 | |
GenBank mRNA | AF155883.1 | - | 4 - 5 | 1295 | 87 | |
AF506002.1 | - | 4 - 5 | 1119 | 87 | ||
AK128964.1 | - | 4 - 5 | 1303 | 87 | ||
AK141265.1 | - | 4 - 5 | 1326 | 87 | ||
BC026392.1 | - | 1 - 2 | 205 | 87 | ||
BC046596.1 | - | 4 - 5 | 1068 | 6312263 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
