Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000007489.mm.1 |
NM_021439.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007489.mm.1 |
NM_021439.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007489.mm.1 |
TC1000000937.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007487.mm.1 |
NM_021439.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007487.mm.1 |
NM_021439.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007487.mm.1 |
TC1000000937.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007486.mm.1 |
NM_021439.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007486.mm.1 |
NM_021439.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007486.mm.1 |
TC1000000937.mm.1 |
MTA TC |