Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100004199.mm.1 |
NM_021510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004199.mm.1 |
NM_021510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004199.mm.1 |
TC1100000544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004194.mm.1 |
NM_021510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004194.mm.1 |
NM_021510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004194.mm.1 |
TC1100000544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004191.mm.1 |
NM_021510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004191.mm.1 |
NM_021510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004191.mm.1 |
TC1100000544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004202.mm.1 |
NM_021510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004202.mm.1 |
NM_021510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004202.mm.1 |
TC1100000544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004193.mm.1 |
NM_021510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004193.mm.1 |
NM_021510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004193.mm.1 |
TC1100000544.mm.1 |
MTA TC |