Assay Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021896.5 | NP_068696.2 | 2 - 3 | 566 | 68 | |
| GenBank mRNA | AF297082.1 | - | 2 - 3 | 541 | 68 | |
| AK004815.1 | - | 2 - 3 | 500 | 68 | ||
| AK028558.1 | - | 2 - 3 | 489 | 68 | ||
| AK031305.1 | - | 2 - 3 | 479 | 68 | ||
| BC057327.1 | - | 2 - 3 | 476 | 6825960 | 68 | |
| BC094310.1 | - | 2 - 3 | 622 | 6838836 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy1a3
Genomic Map