Assay Details
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164533.1 | NP_001158005.1 | 9 - 10 | 1247 | 94 | |
NM_022994.3 | NP_075370.2 | 9 - 10 | 1247 | 94 | ||
GenBank mRNA | AJ250375.1 | - | 9 - 10 | 991 | 94 | |
AK014889.1 | - | 9 - 10 | 917 | 94 | ||
AK021250.1 | - | 4 - 5 | 454 | 94 | ||
AK032275.1 | - | 9 - 10 | 919 | 94 | ||
AK088960.1 | - | 9 - 10 | 917 | 94 | ||
BC019566.1 | - | 9 - 10 | 1247 | 4235517 | 94 | |
CD741520.1 | - | 3 - 4 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22316450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22316451 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975742 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384691 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242695 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958856 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253973 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261469 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525363 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575153 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323790 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866570 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187744 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493088 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78453 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119897 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60329 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73391 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975743 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975742 | - | - |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316466 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22316467 | - | - |
Whole Mouse Genome G4122A | A_52_P292882 | - | - |
Whole Mouse Genome G4122A | A_51_P235580 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129434 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02408434 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02408435 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22316451 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22316450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22316462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03058263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02565657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02585906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02487603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02963980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316464 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22316485 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10499461 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499458 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499455 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499464 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499469 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499459 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499456 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499460 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499463 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499466 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499467 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499462 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499468 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499457 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499465 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450848_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10499454 | - | - |
Mouse Genome 430 2.0 Array | 1450848_at | - | - |
Mouse Genome 430 2.0 Array | 1450848_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65111_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65111_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300017777.mm.1 | NM_001164533.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300017777.mm.1 | NM_001164533.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300017777.mm.1 | NM_022994.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300017777.mm.1 | NM_022994.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300017777.mm.1 | TC0300002329.mm.1 | MTA TC |
Murine Genome U74v2 Set | 94524_PM_at | - | - |
Murine Genome U74v2 Set | 94524_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450848_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450848_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1660528 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1660528 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5420593 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2120039 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002120039 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005420593 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001660528 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722542 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620078 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646230 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2620078 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2722542 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2646230 | - | - |
mouse-6 v1.1 (Illumina) | scl0065111.1_63-S | - | - |
mouse-6 v1.1 (Illumina) | scl065111.1_137-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001977.1_31-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5420593 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1660528 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2120039 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001977.1_31-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl065111.1_137-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0065111.1_63-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120039 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660528 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420593 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2620078 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2722542 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2646230 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2953910 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620078 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2953910 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2646230 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2722542 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl065111.1_137-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001977.1_31-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0065111.1_63-S | - | - |
