Assay Details
Gene Symbol: Dap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022994.3 | NP_075370.2 | 10 - 11 | 1307 | 77 | |
GenBank mRNA | AJ250375.1 | - | 10 - 11 | 1051 | 77 | |
AK014889.1 | - | 10 - 11 | 977 | 77 | ||
AK032275.1 | - | 10 - 11 | 979 | 77 | ||
BC019566.1 | - | 10 - 11 | 1307 | 4235517 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
