Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017770.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017770.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017770.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017770.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017770.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017772.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017772.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017772.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017772.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017772.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017768.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017768.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017768.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017768.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017768.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017773.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017773.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017773.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017773.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017773.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017771.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017771.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017771.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017771.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017771.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017769.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017769.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017769.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017769.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017769.mm.1 |
TC0300002329.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017774.mm.1 |
NM_001164533.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017774.mm.1 |
NM_001164533.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017774.mm.1 |
NM_022994.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017774.mm.1 |
NM_022994.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017774.mm.1 |
TC0300002329.mm.1 |
MTA TC |