Assay Details
| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023040.3 | NP_075527.2 | 2 - 3 | 528 | 73 | |
| GenBank mRNA | AB041561.1 | - | 2 - 3 | 445 | 73 | |
| AF148688.1 | - | 2 - 3 | 308 | 73 | ||
| AK146579.1 | - | 2 - 3 | 529 | 73 | ||
| BC023941.1 | - | 2 - 3 | 528 | 5342526 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of apoptotic process
negative regulation of natural killer cell mediated cytotoxicity
oxidation-reduction process
liver regeneration
negative regulation of oxidative stress-induced neuron death
positive regulation of DNA biosynthetic process
growth factor activity
protein disulfide oxidoreductase activity
oxidoreductase activity
flavin-linked sulfhydryl oxidase activity
thiol oxidase activity
flavin adenine dinucleotide binding
negative regulation of apoptotic process
negative regulation of natural killer cell mediated cytotoxicity
oxidation-reduction process
liver regeneration
negative regulation of oxidative stress-induced neuron death
positive regulation of DNA biosynthetic process
growth factor activity
protein disulfide oxidoreductase activity
oxidoreductase activity
flavin-linked sulfhydryl oxidase activity
thiol oxidase activity
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099062 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144796 | BE691392.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144796 | BI689446.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144796 | NM_023040.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144796 | AB041561.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144796 | BC023941.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35607 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144796 | BE691392.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144796 | BI689446.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144796 | NM_023040.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144796 | AB041561.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144796 | BC023941.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099062 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132791 | - | - |
| Whole Mouse Genome G4122A | A_51_P218805 | NM_023040.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218805 | AB041561.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218805 | AF148688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218805 | AK146579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218805 | BC023941.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140725 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13312253 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380586 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13380581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13526181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13312252 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13526182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31099060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31098984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13379850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10448705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448701 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426995_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452272_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448700 | - | - |
| Mouse Genome 430 2.0 Array | 1426995_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452272_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426995_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452272_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11692_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11692_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014651.mm.1 | NM_023040.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014651.mm.1 | NM_023040.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014651.mm.1 | TC1700001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014652.mm.1 | NM_023040.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014652.mm.1 | NM_023040.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014652.mm.1 | TC1700001637.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426995_a_at | - | - |
| Mouse Expression Set 430 | 1426995_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452272_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452272_a_at | - | - |
| Murine Genome U74v2 Set | 160269_PM_at | - | - |
| Murine Genome U74v2 Set | 160269_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452272_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426995_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426995_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452272_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426995_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452272_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452272_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426995_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 739996 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6450333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006450333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219169 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219169 | - | - |
| mouse-6 v1.1 (Illumina) | scl50203.3_209-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6450333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50203.3_209-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219169 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219169 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50203.3_209-S | - | - |

Gfer
Genomic Map