Assay Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024239.2 | NP_077201.1 | 4 - 5 | 484 | 106 | |
| GenBank mRNA | AB010123.1 | - | 4 - 5 | 431 | 106 | |
| AK017600.1 | - | 4 - 5 | 439 | 106 | ||
| AK019907.1 | - | 4 - 5 | 434 | 106 | ||
| AK046995.1 | - | 4 - 5 | 436 | 106 | ||
| AK136961.1 | - | 4 - 5 | 1035 | 106 | ||
| AK139391.1 | - | 4 - 5 | 459 | 106 | ||
| AK168810.1 | - | 4 - 5 | 457 | 106 | ||
| AK220541.1 | - | 2 - 3 | 6715 | 106 | ||
| BC003497.1 | - | 4 - 5 | 427 | 3153829 | 106 | |
| BC006939.1 | - | 4 - 5 | 482 | 2649718 | 106 | |
| BC025111.1 | - | 4 - 5 | 426 | 4194332 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stambp
Genomic Map