Assay Details
| Gene Symbol: Bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024285.2 | NP_077247.1 | 1 - 2 | 120 | 120 | |
| GenBank mRNA | AF204174.1 | - | 1 - 2 | 118 | 120 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF204174.1,NM_024285.2 transcripts.
Set Membership: |
|

Bves
Genomic Map