Assay Details
| Gene Symbol: Eid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025613.3 | NP_079889.2 | 1 | 543 | 84 | |
| GenBank mRNA | AF509968.1 | - | 1 | 481 | 84 | |
| AK002559.1 | - | 1 | 549 | 84 | ||
| AK005258.1 | - | 1 | 513 | 84 | ||
| AK011293.1 | - | 1 | 535 | 84 | ||
| AK012453.1 | - | 1 | 535 | 84 | ||
| AK014440.1 | - | 1 | 489 | 84 | ||
| AK017543.1 | - | 1 | 519 | 84 | ||
| AK077783.1 | - | 1 | 523 | 84 | ||
| AK078402.1 | - | 1 | 489 | 84 | ||
| AK078409.1 | - | 1 | 489 | 84 | ||
| AK078826.1 | - | 1 | 467 | 84 | ||
| AK148555.1 | - | 1 | 484 | 84 | ||
| AK169604.1 | - | 1 | 517 | 84 | ||
| AK172612.1 | - | 1 | 324 | 84 | ||
| AL357453.1 | - | 1 | 509 | 84 | ||
| BC010712.1 | - | 1 | 528 | 84 | ||
| BC069897.1 | - | 1 | 466 | 84 | ||
| BY703074.1 | - | 1 | 541 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
cell differentiation
regulation of histone acetylation
negative regulation of transcription, DNA-templated
regulation of catalytic activity
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
protein binding
histone acetyltransferase regulator activity
histone acetyltransferase binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
cell differentiation
regulation of histone acetylation
negative regulation of transcription, DNA-templated
regulation of catalytic activity
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
protein binding
histone acetyltransferase regulator activity
histone acetyltransferase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604060 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097022 | BU610167.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097022 | BU700648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097022 | NM_025613.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097017 | AL357452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097017 | AL357453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097017 | BC010712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097017 | BC069897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097017 | AK148555.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123835 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097022 | BU610167.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097022 | BU700648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097022 | NM_025613.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097017 | AL357452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097017 | AL357453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097017 | BC010712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097017 | BC069897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097017 | AK148555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205564 | AL357452.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205564 | AL357453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205564 | AK148555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205564 | AK172612.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205564 | AK017543.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21604026 | - | - |
| Whole Mouse Genome G4122A | A_51_P205564 | AL357452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205564 | AL357453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205564 | AK148555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205564 | AK172612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205564 | AK017543.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130118 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01672137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01951209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01672142 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01672141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21604060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21603992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21604029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01691372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01183581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01376759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21604059 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10475626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475628 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448405_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416614_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448406_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10475625 | - | - |
| Mouse Genome 430 2.0 Array | 1448406_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448406_at | - | - |
| Mouse Genome 430 2.0 Array | 1448405_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416614_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416614_at | - | - |
| Mouse Genome 430 2.0 Array | 1448405_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58521_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58521_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200014687.mm.1 | NM_025613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014687.mm.1 | NM_025613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014687.mm.1 | TC0200001886.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200014685.mm.1 | NM_025613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014685.mm.1 | NM_025613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014685.mm.1 | TC0200001886.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200014686.mm.1 | NM_025613.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014686.mm.1 | NM_025613.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014686.mm.1 | TC0200001886.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448405_a_at | - | - |
| Mouse Expression Set 430 | 1448405_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448406_at | - | - |
| Mouse Expression Set 430 | 1416614_at | - | - |
| Mouse Expression Set 430 | 1416614_PM_at | - | - |
| Mouse Expression Set 430 | 1448406_PM_at | - | - |
| Murine Genome U74v2 Set | 99191_PM_at | - | - |
| Murine Genome U74v2 Set | 99191_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448405_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416614_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416614_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448405_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448406_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448406_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448405_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448406_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416614_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448406_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416614_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448405_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 909343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6620132 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620132 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2669766 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2669766 | - | - |
| mouse-6 v1.1 (Illumina) | scl20341.2.61_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620132 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20341.2.61_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620132 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2669766 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2669766 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20341.2.61_4-S | - | - |

Eid1
Genomic Map