Assay Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012324.1 | - | 1 - 2 | 115 | 105 | |
| AK078925.1 | - | 1 - 2 | 127 | 105 | ||
| AK079385.1 | - | 1 - 2 | 123 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012324.1, AK078925.1, AK079385.1 transcripts.
Set Membership: |
|

Bcs1l
Genomic Map