Assay Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025816.3 | NP_080092.2 | 5 - 6 | 744 | 105 | |
| GenBank mRNA | AK004574.1 | - | 5 - 6 | 726 | 105 | |
| AK146076.1 | - | 5 - 6 | 724 | 105 | ||
| BC014798.1 | - | 5 - 6 | 665 | 3962810 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tax1bp1
Genomic Map