Assay Details
| Gene Symbol: Echdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110195.1 | NP_001103665.1 | 3 - 4 | 494 | 119 | |
| NM_025855.4 | NP_080131.4 | 3 - 4 | 596 | 119 | ||
| GenBank mRNA | AK006444.1 | - | 3 - 4 | 473 | 119 | |
| AK028775.1 | - | 3 - 4 | 493 | 119 | ||
| AK145162.1 | - | 556 | 119 | |||
| AK150932.1 | - | 3 - 4 | 478 | 119 | ||
| AK152285.1 | - | 3 - 4 | 478 | 119 | ||
| AK153454.1 | - | 3 - 4 | 478 | 119 | ||
| AK166589.1 | - | 3 - 4 | 518 | 119 | ||
| AK166634.1 | - | 3 - 4 | 518 | 119 | ||
| AK166660.1 | - | 3 - 4 | 518 | 119 | ||
| BC066183.1 | - | 3 - 4 | 461 | 30473683 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Echdc1
Genomic Map