Assay Details
| Gene Symbol: Ifrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025903.2 | NP_080179.1 | 9 - 10 | 1188 | 69 | |
| GenBank mRNA | AK002702.1 | - | 3 - 4 | 413 | 69 | |
| AK007687.1 | - | 9 - 10 | 1189 | 69 | ||
| AK149496.1 | - | 9 - 10 | 1169 | 69 | ||
| AK156625.1 | - | 8 - 9 | 1264 | 69 | ||
| AK165967.1 | - | 9 - 10 | 1182 | 69 | ||
| AK166130.1 | - | 9 - 10 | 1256 | 69 | ||
| AK166258.1 | - | 9 - 10 | 1146 | 69 | ||
| AY298860.1 | - | 9 - 10 | 1017 | 69 | ||
| BC023486.1 | - | 9 - 10 | 1135 | 69 | ||
| BC037434.1 | - | 9 - 10 | 1144 | 2651172 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd2
Genomic Map