Assay Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026106.4 | NP_080382.2 | 2 - 3 | 1076 | 90 | |
| GenBank mRNA | AK007230.1 | - | 2 - 3 | 1051 | 90 | |
| AK018040.1 | - | 2 - 3 | 999 | 90 | ||
| AK031057.1 | - | 2 - 3 | 1010 | 90 | ||
| AK075713.1 | - | 2 - 3 | 1076 | 90 | ||
| AK090113.1 | - | 2 - 3 | 1058 | 90 | ||
| AK139866.1 | - | 2 - 3 | 1006 | 90 | ||
| AK144749.1 | - | 2 - 3 | 1070 | 90 | ||
| AK146573.1 | - | 2 - 3 | 1039 | 90 | ||
| BC013461.1 | - | 2 - 3 | 645 | 4222118 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970972 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8879 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173895 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109477 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498922 | BC013461.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498922 | AK144749.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498922 | AK146573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498922 | AK090113.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498922 | AK139866.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23970963 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173895 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109477 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01183677 | - | - |
| Whole Mouse Genome G4122A | A_51_P498922 | BC013461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498922 | AK144749.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498922 | AK146573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498922 | AK090113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498922 | AK139866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | BC013461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK144749.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK146573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK090113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P462350 | AK031057.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173895 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109477 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04455548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04455544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04455553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04680666 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04455549 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04455571 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23970976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23970992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23971025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04465274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04455571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04778703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04455545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04347988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23971005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23970943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04276998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04859854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523958 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416018_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416019_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523955 | - | - |
| Mouse Genome 430 2.0 Array | 1416018_at | - | - |
| Mouse Genome 430 2.0 Array | 1416019_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416019_at | - | - |
| Mouse Genome 430 2.0 Array | 1416018_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13486_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13486_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500009329.mm.1 | TC0500001098.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009329.mm.1 | NM_026106.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009329.mm.1 | NM_026106.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009330.mm.1 | TC0500001098.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009330.mm.1 | NM_026106.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009330.mm.1 | NM_026106.4 | Refseq |
| Mouse Expression Set 430 | 1416019_at | - | - |
| Mouse Expression Set 430 | 1416019_PM_at | - | - |
| Mouse Expression Set 430 | 1416018_at | - | - |
| Mouse Expression Set 430 | 1416018_PM_at | - | - |
| Murine Genome U74v2 Set | 160449_PM_at | - | - |
| Murine Genome U74v2 Set | 160449_at | - | - |
| Murine Genome U74v2 Set | 168968_r_at | - | - |
| Murine Genome U74v2 Set | 168968_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416019_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416019_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416018_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416018_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416018_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416019_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416019_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416018_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 856453 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5860750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5860750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860750 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2728290 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2728290 | - | - |
| mouse-6 v1.1 (Illumina) | scl27468.4_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860750 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27468.4_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860750 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2728290 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2728290 | - | - |

Dr1
Genomic Map