Assay Details
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026538.3 | NP_080814.1 | 2 - 3 | 230 | 60 | |
GenBank mRNA | AK011136.1 | - | 2 - 3 | 240 | 60 | |
AK076047.1 | - | 2 - 3 | 248 | 60 | ||
AK076975.1 | - | 2 - 3 | 230 | 60 | ||
AK077514.1 | - | 2 - 3 | 243 | 60 | ||
AK145628.1 | - | 2 - 3 | 271 | 60 | ||
AK159793.1 | - | 2 - 3 | 234 | 60 | ||
AK169654.1 | - | 2 - 3 | 245 | 60 | ||
BC018291.1 | - | 2 - 3 | 224 | 4190167 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
