Assay Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026623.3 | NP_080899.1 | 5 - 6 | 698 | 130 | |
| GenBank mRNA | AK011688.2 | - | 5 - 6 | 688 | 130 | |
| AK012308.1 | - | 5 - 6 | 130 | |||
| AK019433.1 | - | 5 - 6 | 681 | 130 | ||
| AK020150.1 | - | 5 - 6 | 679 | 130 | ||
| AK146419.1 | - | 5 - 6 | 664 | 130 | ||
| AK147061.1 | - | 681 | 130 | |||
| AK160147.1 | - | 5 - 6 | 664 | 130 | ||
| AK208594.1 | - | 4 - 5 | 273 | 130 | ||
| BC008270.1 | - | 5 - 6 | 681 | 2811199 | 130 | |
| BC090834.1 | - | 657 | 6855233 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt21
Genomic Map