Assay Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026819.3 | NP_081095.2 | 3 - 4 | 563 | 74 | |
| GenBank mRNA | AK003958.1 | - | 3 - 4 | 453 | 74 | |
| AK146317.1 | - | 3 - 4 | 510 | 74 | ||
| AK159964.1 | - | 3 - 4 | 485 | 74 | ||
| AK168115.1 | - | 3 - 4 | 510 | 74 | ||
| AK168147.1 | - | 3 - 4 | 510 | 74 | ||
| BC003930.1 | - | 3 - 4 | 494 | 3485584 | 74 | |
| BC023825.1 | - | 2 - 3 | 1605 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs1
Genomic Map