Assay Details
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026819.3 | NP_081095.2 | 7 - 8 | 985 | 69 | |
GenBank mRNA | AK003958.1 | - | 7 - 8 | 875 | 69 | |
AK146317.1 | - | 7 - 8 | 932 | 69 | ||
AK159964.1 | - | 7 - 8 | 907 | 69 | ||
AK168115.1 | - | 7 - 8 | 932 | 69 | ||
AK168147.1 | - | 7 - 8 | 932 | 69 | ||
BC003930.1 | - | 7 - 8 | 916 | 3485584 | 69 | |
BC023825.1 | - | 6 - 7 | 2026 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
