Assay Details
| Gene Symbol: Cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029976.3 | NP_084252.1 | 2 - 3 | 434 | 76 | |
| GenBank mRNA | AB041653.1 | - | 2 - 3 | 402 | 76 | |
| AK020751.1 | - | 2 - 3 | 407 | 76 | ||
| AK077515.1 | - | 2 - 3 | 259 | 76 | ||
| AK150573.1 | - | 2 - 3 | 390 | 76 | ||
| AK171740.1 | - | 2 - 3 | 369 | 76 | ||
| BC099516.1 | - | 2 - 3 | 165 | 76 | ||
| BC116362.2 | - | 2 - 3 | 394 | 40099997 | 76 | |
| BC116363.2 | - | 2 - 3 | 394 | 40099998 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796700 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796702 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180406 | BC116363.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180406 | BC116362.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180406 | AK150573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180406 | AK171740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180406 | BC099516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506312 | BQ032176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506312 | AK150573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506312 | AK171740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506312 | BC099516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P506312 | NM_029976.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142362 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149458 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506312 | BQ032176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506312 | AK150573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506312 | AK171740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506312 | BC099516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P506312 | NM_029976.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180406 | BC116363.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180406 | BC116362.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180406 | AK150573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180406 | AK171740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180406 | BC099516.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27796676 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149458 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07341728 | - | - |
| Whole Mouse Genome G4122A | A_52_P284658 | BC116363.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284658 | BC116362.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284658 | AK150573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284658 | AK171740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284658 | BC099516.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130282 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149458 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09329590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09521767 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09521770 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09329589 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09521766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27796700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27796800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09657301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09521770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09005331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09521769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09501285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09329589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09638795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09336151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09638794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09521756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09502989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27796731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09127977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375907 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375908 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440715_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452200_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375903 | - | - |
| Mouse Genome 430 2.0 Array | 1452200_at | - | - |
| Mouse Genome 430 2.0 Array | 1452200_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440715_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1440715_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52626_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52626_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100004522.mm.1 | NM_029976.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004522.mm.1 | NM_029976.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004522.mm.1 | TC1100000582.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004523.mm.1 | NM_029976.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004523.mm.1 | NM_029976.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004523.mm.1 | TC1100000582.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100004524.mm.1 | NM_029976.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004524.mm.1 | NM_029976.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100004524.mm.1 | TC1100000582.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452200_PM_at | - | - |
| Mouse Expression Set 430 | 1452200_at | - | - |
| Mouse Expression Set 430 | 1440715_PM_s_at | - | - |
| Mouse Expression Set 430 | 1440715_s_at | - | - |
| Murine Genome U74v2 Set | 160125_at | - | - |
| Murine Genome U74v2 Set | 160125_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452200_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452200_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452200_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452200_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440715_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440715_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7050603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236906 | - | - |
| mouse-6 v1.1 (Illumina) | scl41586.4_460-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41586.4_460-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1236906 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236906 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41586.4_460-S | - | - |

Cdkn2aipnl
Genomic Map