Assay Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030113.2 | NP_084389.2 | 4 - 5 | 503 | 121 | |
| GenBank mRNA | AF297029.1 | - | 4 - 5 | 407 | 121 | |
| AF297030.1 | - | 4 - 5 | 466 | 121 | ||
| AY179965.1 | - | 4 - 5 | 379 | 121 | ||
| AY179966.1 | - | 4 - 5 | 379 | 121 | ||
| AY179967.1 | - | 4 - 5 | 379 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap10
Genomic Map