Assay Details
Gene Symbol: Mrgprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030726.1 | NP_109651.1 | 1 | 1761 | 71 | |
GenBank mRNA | AF295365.1 | - | 1 | 1761 | 71 | |
BC116696.1 | - | 1 | 1391 | 40105568 | 71 | |
BC117525.1 | - | 1 | 1389 | 40105567 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
