Assay Details
Gene Symbol: Cacnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159320.2 | NP_001152792.1 | 6 - 7 | 713 | 62 | |
NM_001282977.1 | NP_001269906.1 | 7 - 8 | 986 | 62 | ||
NM_031173.4 | NP_112450.1 | 7 - 8 | 986 | 62 | ||
GenBank mRNA | AF322905.1 | - | 7 - 8 | 990 | 62 | |
AI466668.1 | - | 2 - 3 | 217 | 62 | ||
AK036404.1 | - | 6 - 7 | 714 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
