Assay Details
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031402.2 | NP_113579.2 | 7 - 8 | 1276 | 103 | |
GenBank mRNA | AF329198.1 | - | 7 - 8 | 1273 | 103 | |
AK132405.1 | - | 7 - 8 | 1014 | 103 | ||
AK133870.1 | - | 7 - 8 | 1277 | 103 | ||
AK134273.1 | - | 7 - 8 | 1212 | 103 | ||
AK137611.1 | - | 7 - 8 | 1277 | 103 | ||
BC040768.1 | - | 7 - 8 | 1605 | 4458200 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
