Assay Details
Gene Symbol: Rnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053113.2 | NP_444343.2 | 2 | 382 | 87 | |
GenBank mRNA | BC020070.1 | - | 2 | 381 | 3982432 | 87 |
BC027557.1 | - | 2 | 382 | 1445853 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455338 | BC027557.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P455338 | BC027557.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170668 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455338 | BC027557.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P455338 | BC027557.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600811 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600825 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600804 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600827 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170668 | - | - |
Whole Mouse Genome G4122A | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455338 | BC027557.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455338 | NM_053113.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P455338 | BC027557.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119405 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170668 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11271887 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11271874 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11292631 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11271876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11271876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11271878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600780 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11798519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11798518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11482026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11373552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11292631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600786 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10419157 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10419569 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10419155 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425295_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10419568 | - | - |
Mouse Genome 430 2.0 Array | 1425295_at | - | - |
Mouse Genome 430 2.0 Array | 1425295_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93726_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93726_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400015046.mm.1 | NM_053113.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400015046.mm.1 | NM_053113.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400015046.mm.1 | TC1400002002.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400015047.mm.1 | NM_053113.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400015047.mm.1 | NM_053113.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400015047.mm.1 | TC1400002002.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425295_PM_at | - | - |
Mouse Expression Set 430 | 1425295_at | - | - |
Mouse Genome 430A 2.0 Array | 1425295_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425295_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425295_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425295_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 586136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2260292 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260292 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652359 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2652359 | - | - |
mouse-6 v1.1 (Illumina) | scl45605.2.1_15-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260292 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl45605.2.1_15-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260292 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2890019 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2890019 | - | - |
