Assay Details
| Gene Symbol: Cntnap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130457.2 | NP_569724.2 | 17 - 18 | 2852 | 76 | |
| GenBank mRNA | AF333770.1 | - | 17 - 18 | 2817 | 76 | |
| AK081891.1 | - | 17 - 18 | 2845 | 76 | ||
| AK142816.1 | - | 17 - 18 | 2848 | 76 | ||
| AK173255.1 | - | 17 - 18 | 2827 | 76 | ||
| BC034628.1 | - | 17 - 18 | 2852 | 5369648 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntnap4
Genomic Map