Assay Details
| Gene Symbol: Krtap19-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130873.1 | NP_570943.1 | 1 | 266 | 86 | |
| GenBank mRNA | AF345294.1 | - | 1 | 266 | 86 | |
| BC116200.1 | - | 1 | 266 | 40090087 | 86 | |
| BC116201.1 | - | 1 | 266 | 40090088 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151209 | NM_130873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151209 | AF345294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151209 | BC116200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151209 | BC116201.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157079 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151209 | NM_130873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151209 | AF345294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151209 | BC116200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151209 | BC116201.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30948556 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157079 | - | - |
| Whole Mouse Genome G4122A | A_51_P360422 | NM_130873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360422 | AF345294.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136976 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681723 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681713 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681704 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12681710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30948501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12752247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12681723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12603328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12862084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12662628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30948549 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10440692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440693 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426203_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440690 | - | - |
| Mouse Genome 430 2.0 Array | 1426203_at | - | - |
| Mouse Genome 430 2.0 Array | 1426203_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170654_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170654_PM_at | - | - |
| Mouse Expression Set 430 | 1426203_at | - | - |
| Mouse Expression Set 430 | 1426203_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426203_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426203_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426203_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426203_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 473762 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6130563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2645563 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2645563 | - | - |
| mouse-6 v1.1 (Illumina) | scl48237.1.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48237.1.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2645563 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2645563 | - | - |

Krtap19-4
Genomic Map