Assay Details
| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133193.3 | NP_573456.1 | 3 - 4 | 496 | 73 | |
| GenBank mRNA | AF284433.1 | - | 3 - 4 | 501 | 73 | |
| AK054530.1 | - | 1 - 2 | 2997 | 73 | ||
| AK085780.1 | - | 3 - 4 | 364 | 73 | ||
| BC004628.1 | - | 3 - 4 | 490 | 73 | ||
| BC117797.2 | - | 3 - 4 | 497 | 40092208 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl2
Genomic Map