Assay Details
| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133193.3 | NP_573456.1 | 5 - 6 | 860 | 80 | |
| GenBank mRNA | AF284433.1 | - | 5 - 6 | 865 | 80 | |
| AK085780.1 | - | 5 - 6 | 728 | 80 | ||
| BC117797.2 | - | 5 - 6 | 861 | 40092208 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl2
Genomic Map