Assay Details
| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133193.3 | NP_573456.1 | 9 - 10 | 1339 | 108 | |
| GenBank mRNA | AF284433.1 | - | 9 - 10 | 1344 | 108 | |
| AK085780.1 | - | 9 - 10 | 1207 | 108 | ||
| BC117797.2 | - | 9 - 10 | 1340 | 40092208 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl2
Genomic Map