Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
TC0500001338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_001177594.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_001177594.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_001177595.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_001177595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_133221.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500011668.mm.1 |
NM_133221.2 |
Refseq |