| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600022374.mm.1 |
TC0600002917.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022374.mm.1 |
NM_133926.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022374.mm.1 |
NM_133926.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022380.mm.1 |
TC0600002917.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022380.mm.1 |
NM_133926.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022380.mm.1 |
NM_133926.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022379.mm.1 |
TC0600002917.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600022379.mm.1 |
NM_133926.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600022379.mm.1 |
NM_133926.2 |
Refseq |