Assay Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133930.1 | NP_598691.1 | 4 - 5 | 792 | 62 | |
| GenBank mRNA | AB009020.1 | - | 4 - 5 | 797 | 62 | |
| AK077726.1 | - | 4 - 5 | 936 | 62 | ||
| AK155777.1 | - | 4 - 5 | 802 | 62 | ||
| AK182708.1 | - | 2 - 3 | 147 | 62 | ||
| AK194808.1 | - | 2 - 3 | 140 | 62 | ||
| BC010804.1 | - | 4 - 5 | 792 | 4239786 | 62 | |
| BC023893.1 | - | 4 - 5 | 803 | 5310111 | 62 | |
| BC025932.1 | - | 4 - 5 | 698 | 5102571 | 62 | |
| BC029065.1 | - | 4 - 5 | 828 | 4935296 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creld1
Genomic Map