Assay Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133930.1 | NP_598691.1 | 9 - 10 | 1384 | 60 | |
| GenBank mRNA | AB009020.1 | - | 9 - 10 | 1389 | 60 | |
| AK050160.1 | - | 2 - 3 | 165 | 60 | ||
| AK050455.1 | - | 2 - 3 | 165 | 60 | ||
| AK077726.1 | - | 9 - 10 | 1529 | 60 | ||
| AK155777.1 | - | 9 - 10 | 1394 | 60 | ||
| BC010804.1 | - | 9 - 10 | 1384 | 4239786 | 60 | |
| BC023893.1 | - | 9 - 10 | 1395 | 5310111 | 60 | |
| BC025932.1 | - | 9 - 10 | 1290 | 5102571 | 60 | |
| BC029065.1 | - | 9 - 10 | 1420 | 4935296 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759773 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111492 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162964 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24759791 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01421237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162964 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06031171 | - | - |
| Whole Mouse Genome G4122A | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC025932.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC029065.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC010804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC023058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205545 | BC023893.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121820 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01421237 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162964 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05585504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05585519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05585515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05585510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24759773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05318335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05318337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05657674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05527237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05527227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05662320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05602879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05559392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05602873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05318343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24759792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05585504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05602872 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540736 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416537_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540727 | - | - |
| Mouse Genome 430 2.0 Array | 1416537_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416537_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171508_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 171508_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600009812.mm.1 | TC0600001253.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009812.mm.1 | NM_133930.2 | Refseq |
| Mouse Expression Set 430 | 1416537_at | - | - |
| Mouse Expression Set 430 | 1416537_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416537_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416537_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416537_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416537_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102190427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2060253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102190427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232170 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254175 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232170 | - | - |
| mouse-6 v1.1 (Illumina) | scl29631.10_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102190427 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29631.10_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102190427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232170 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254175 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232170 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29631.10_10-S | - | - |

Creld1
Genomic Map