Assay Details
| Gene Symbol: Psmg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134138.1 | NP_598899.1 | 3 - 4 | 365 | 72 | |
| GenBank mRNA | AB031387.1 | - | 3 - 4 | 365 | 72 | |
| AK077569.1 | - | 3 - 4 | 352 | 72 | ||
| AK167840.1 | - | 3 - 4 | 606 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmg2
Genomic Map