Assay Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127338.1 | NP_001120810.1 | 4 - 5 | 509 | 71 | |
| NM_138600.4 | NP_613066.2 | 2 - 3 | 315 | 71 | ||
| GenBank mRNA | AK004991.1 | - | 4 - 5 | 443 | 71 | |
| AK145873.1 | - | 2 - 3 | 258 | 71 | ||
| AK160117.1 | - | 2 - 3 | 242 | 71 | ||
| AK169195.1 | - | 2 - 3 | 258 | 71 | ||
| AK187582.1 | - | 1 - 2 | 58 | 71 | ||
| BC012407.1 | - | 2 - 3 | 277 | 4191228 | 71 | |
| BY017676.1 | - | 2 - 3 | 315 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh7a1
Genomic Map