Assay Details
| Gene Symbol: Ccdc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138603.3 | NP_613069.3 | 4 - 5 | 599 | 80 | |
| GenBank mRNA | AF229637.1 | - | 4 - 5 | 540 | 80 | |
| AK010536.1 | - | 4 - 5 | 517 | 80 | ||
| AK039573.1 | - | 4 - 5 | 572 | 80 | ||
| BC011195.1 | - | 4 - 5 | 504 | 4188459 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc22
Genomic Map