Assay Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285463.1 | NP_001272392.1 | 11 - 12 | 1218 | 148 | |
| NM_001285473.1 | NP_001272402.1 | 7 - 8 | 808 | 148 | ||
| NM_139150.5 | NP_631889.1 | 9 - 10 | 1091 | 148 | ||
| GenBank mRNA | AB221600.1 | - | 8 - 9 | 874 | 148 | |
| AF454947.1 | - | 9 - 10 | 1060 | 148 | ||
| AK044738.1 | - | 11 - 12 | 1219 | 148 | ||
| BC138644.1 | - | 8 - 9 | 997 | 8861664 | 148 | |
| BC138645.1 | - | 8 - 9 | 997 | 8861665 | 148 | |
| BC145350.1 | - | 8 - 9 | 993 | 148 | ||
| EU170502.1 | - | 7 - 8 | 789 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carf
Genomic Map