Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100003813.mm.1 |
NM_001285463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003813.mm.1 |
NM_001285473.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003813.mm.1 |
NM_139150.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003813.mm.1 |
NR_104344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003813.mm.1 |
TC0100000465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003814.mm.1 |
NM_001285463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003814.mm.1 |
NM_001285473.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003814.mm.1 |
NM_139150.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003814.mm.1 |
NR_104344.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003814.mm.1 |
TC0100000465.mm.1 |
MTA TC |