Assay Details
| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 14 - 15 | 2111 | 81 | |
| GenBank mRNA | AB076696.1 | - | 14 - 15 | 2111 | 81 | |
| AK085882.1 | - | 14 - 15 | 1892 | 81 | ||
| AK129032.1 | - | 14 - 15 | 1857 | 81 | ||
| AK135820.1 | - | 14 - 15 | 1918 | 81 | ||
| AK147955.1 | - | 14 - 15 | 1881 | 81 | ||
| BC027273.1 | - | 14 - 15 | 1832 | 81 | ||
| BC043924.1 | - | 6 - 7 | 737 | 81 | ||
| BC058087.1 | - | 14 - 15 | 1845 | 6816660 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap5
Genomic Map